missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Polyethylene sheet, Low Density, 6.35mm (0.25 in.) thick

$38.95 - $38.95

Chemical Identifiers

CAS 9002-88-4
MDL Number MFCD00084423
Chemical Name or Material Polyethylene sheet, Low Density, 6.35mm (0.25 in.) thick
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA45197HB SDS Alfa Aesar™
45197HB
300x300mm Each for $38.95
Specifications Description & Specifications

Chemical Identifiers

9002-88-4
MFCD00084423
Polyethylene sheet, Low Density, 6.35mm (0.25 in.) thick

Specifications

Odorless
9002-88-4
Polyethylene sheet, Low Density, 6.35mm (0.25 in.) thick
MFCD00084423
300x300mm

Safety and Handling

TSCA : TSCA