Please login to your online account to display your discounted pricing

Alfa Aesar™ Polyethylene sheet, Low Density, 3.18mm (0.125 in.) thick

$18.69 - $18.69

Chemical Identifiers

Chemical Name or Material Polyethylene sheet
CAS 9002-88-4
Name Note Low Density, 3.18mm (0.125 in.) thick
MDL Number MFCD00084423
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA45175HB

  SDS
alfa aesar™
45175HB
300x300mm Each for $18.69
Description & Specifications

Specifications

Density 0.92g/mL
TSCA TSCA
MDL Number MFCD00084423
Physical Form Sheet
Color White
Chemical Name or Material Polyethylene sheet
Odor Odorless
CAS 9002-88-4
Name Note Low Density, 3.18mm (0.125 in.) thick
Quantity 300x300mm