Please login to your online account to display your discounted pricing

Alfa Aesar™ Polyethylene sheet, Low Density, 12.7mm (0.5 in.) thick

$54.65 - $54.65

Chemical Identifiers

Chemical Name or Material Polyethylene sheet
CAS 9002-88-4
Name Note Low Density, 12.7mm (0.5 in.) thick
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA45216HB

  SDS
alfa aesar™
45216HB
300x300mm Each for $54.65
Description & Specifications

Specifications

Chemical Name or Material Polyethylene sheet
TSCA TSCA
Name Note Low Density, 12.7mm (0.5 in.) thick
CAS 9002-88-4
Odor Odorless
Quantity 300x300mm
RTECS Number TQ3325000