Please login to your online account to display your discounted pricing

Alfa Aesar™ Polyethylene rod, Low Density, 6.35mm (0.25 in.) dia.

$17.77 - $17.77

Chemical Identifiers

Chemical Name or Material Polyethylene rod
Name Note Low Density, 6.35mm (0.25 in.) dia.
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA45158BV

  SDS
alfa aesar™
45158BV
30cm Each for $17.77
Description & Specifications

Specifications

Name Note Low Density, 6.35mm (0.25 in.) dia.
Quantity 30cm
Physical Form Rod
Odor Odorless
TSCA TSCA
Chemical Name or Material Polyethylene rod