Please login to your online account to display your discounted pricing

Alfa Aesar™ Polyethylene rod, Low Density, 19mm (0.75 in.) dia.

$44.00 - $44.00

Chemical Identifiers

Chemical Name or Material Polyethylene rod
CAS 9002-88-4
Name Note Low Density, 19mm (0.75 in.) dia.
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA45219BV

  SDS
alfa aesar™
45219BV
30cm Each for $44.00
Description & Specifications

Specifications

Chemical Name or Material Polyethylene rod
Name Note Low Density, 19mm (0.75 in.) dia.
Physical Form Rod
TSCA TSCA
CAS 9002-88-4
Odor Odorless
Quantity 30cm