Please login to your online account to display your discounted pricing

Alfa Aesar™ Polyethylene rod, Low Density, 12.7mm (0.5 in.) dia.

$35.55 - $35.55

Chemical Identifiers

Chemical Name or Material Polyethylene rod
Name Note Low Density, 12.7mm (0.5 in.) dia.
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA45179BV

  SDS
alfa aesar™
45179BV
30cm Each for $35.55
Description & Specifications

Specifications

Name Note Low Density, 12.7mm (0.5 in.) dia.
Physical Form Rod
Quantity 30cm
Chemical Name or Material Polyethylene rod
Odor Odorless
TSCA TSCA