missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Polyethylene rod, High Density, 19mm (0.75 in.) dia. 2.5cm, Alfa Aesar™

Manufacturer:  Alfa Aesar 451801Z

 View more versions of this product

Catalog No. AA451801Z

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Polyethylene rod
9002-88-4
MFCD00084423
VGGSQFUCUMXWEO-UHFFFAOYSA-N
ethene
6325
Rod
2.5cm
High Density, 19mm (0.75 in.) dia.
C2H4
ethylene, acetene, elayl, olefiant gas, athylen, etileno, liquid ethylene, polyethylene, bicarburretted hydrogen, plastipore
C=C
28.054
CHEBI:18153
Odorless

Chemical Identifiers

9002-88-4
28.054
VGGSQFUCUMXWEO-UHFFFAOYSA-N
6325
ethene
C2H4
MFCD00084423
ethylene, acetene, elayl, olefiant gas, athylen, etileno, liquid ethylene, polyethylene, bicarburretted hydrogen, plastipore
CHEBI:18153
C=C
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents