Please login to your online account to display your discounted pricing

Alfa Aesar™ Polyethylene rod, High Density, 12.7mm (0.5 in.) dia.

$17.77 - $17.77

Chemical Identifiers

Chemical Name or Material Polyethylene rod
Name Note High Density, 12.7mm (0.5 in.) dia.
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA45220BV

  SDS
alfa aesar™
45220BV
30cm Each for $17.77
Description & Specifications

Specifications

Odor Odorless
Chemical Name or Material Polyethylene rod
TSCA TSCA
Quantity 30cm
Name Note High Density, 12.7mm (0.5 in.) dia.
Physical Form Rod