Learn More
Polyethylene powder, low density, 500 micron, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1023922
Description
Polyethylene powder is used as an additive to unsaturated polyesters, epoxides and other polymers to impart the unique properties of ultra-high-molecular-weight polyethylene (UHMWPE). It is widely used for manufacturing various containers, trays, dispensing bottles, plastic wraps, plastic bags for computer components and various molded laboratory equipment.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Polyethylene powder | |
Powder | |
(C2H4)n | |
0.92 | |
ethylene, acetene, elayl, olefiant gas, athylen, etileno, liquid ethylene, polyethylene, bicarburretted hydrogen, plastipore | |
100 g | |
*-CC-* | |
28.05 | |
CHEBI:18153 |
Low density, 500 micron | |
9002-88-4 | |
MFCD00084423 | |
14,7567 | |
Insoluble in water. | |
VGGSQFUCUMXWEO-UHFFFAOYSA-N | |
ethene | |
6325 | |
Odorless |
Chemical Identifiers
9002-88-4 | |
28.05 | |
VGGSQFUCUMXWEO-UHFFFAOYSA-N | |
6325 | |
ethene |
(C2H4)n | |
MFCD00084423 | |
ethylene, acetene, elayl, olefiant gas, athylen, etileno, liquid ethylene, polyethylene, bicarburretted hydrogen, plastipore | |
CHEBI:18153 | |
*-CC-* |
Safety and Handling
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only