missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Polyethylene oxide, M.W. >5,000,000, Thermo Scientific Chemicals
$202.92 - $802.70
Chemical Identifiers
CAS | 25322-68-3 |
---|---|
Molecular Formula | (C2H4O)n |
Molecular Weight (g/mol) | 62.07 |
MDL Number | MFCD01779601 |
InChI Key | LYCAIKOWRPUZTN-UHFFFAOYSA-N |
IUPAC Name | ethane-1,2-diol |
SMILES | [H]OCCO |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1553622
|
Thermo Scientific Chemicals
A1553622 |
100 g |
Each for $202.92
|
|
|||||
AAA1553636
|
Thermo Scientific Chemicals
A1553636 |
500 g |
Each for $802.70
|
|
|||||
Description
It is used as a thickening agent and gel binder in the pharmaceutical industry. It is used as a analytical reagents, substrates, flocculants, dispersants, lubricants, emulsifiers, moisturizers, food additives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
25322-68-3 | |
62.07 | |
LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
[H]OCCO |
(C2H4O)n | |
MFCD01779601 | |
ethane-1,2-diol |
Specifications
5 to 7 | |
∼175°C | |
MFCD01779601 | |
100 g | |
[H]OCCO | |
62.07 | |
Odorless |
25322-68-3 | |
(C2H4O)n | |
Soluble in water. | |
LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
ethane-1,2-diol | |
>5,000,000 | |
Polyethylene oxide |
Safety and Handling
EINECSNumber : 500-038-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only