Please login to your online account to display your discounted pricing

Alfa Aesar™ Polyethylene, medium density

$19.45 - $164.24

Chemical Identifiers

Chemical Name or Material Polyethylene
CAS 9002-88-4
Name Note Medium density
Molecular Formula (CH2CH2)n
MDL Number MFCD00084423
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4508518

  SDS
alfa aesar™
4508518
50g Each for $19.45

AA4508530

  SDS
alfa aesar™
4508530
250g Each for $57.57

AA45085A1

  SDS
alfa aesar™
45085A1
1kg Each for $164.24
Description & Specifications

Specifications

Odor Odorless
Chemical Name or Material Polyethylene
Quantity 50g
Name Note Medium density
MDL Number MFCD00084423
CAS 9002-88-4
Density 0.94g/mL
Molecular Formula (CH2CH2)n
TSCA TSCA