missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Polyethylene, medium density

Manufacturer:  Alfa Aesar™ 4508530

 View more versions of this product

Catalog No. AA4508530



Specifications

Specifications

Polyethylene
9002-88-4
C2H4
ethylene, acetene, elayl, olefiant gas, athylen, etileno, liquid ethylene, polyethylene, bicarburretted hydrogen, plastipore
C=C
28.054
CHEBI:18153
250g
Medium density
0.94g/mL
MFCD00084423
VGGSQFUCUMXWEO-UHFFFAOYSA-N
ethene
6325
Odorless

Chemical Identifiers

9002-88-4
28.054
VGGSQFUCUMXWEO-UHFFFAOYSA-N
6325
ethene
C2H4
MFCD00084423
ethylene, acetene, elayl, olefiant gas, athylen, etileno, liquid ethylene, polyethylene, bicarburretted hydrogen, plastipore
CHEBI:18153
C=C
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents