Please login to your online account to display your discounted pricing

Alfa Aesar™ Polyethylene, high density

$43.73 - $136.23

Chemical Identifiers

Chemical Name or Material Polyethylene
CAS 9002-88-4
Name Note High density
MDL Number MFCD00084423
Merck Index 14,7567
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4173130

  SDS
alfa aesar™
4173130
250g Each for $43.73

AA41731A1

  SDS
alfa aesar™
41731A1
1kg Each for $136.23
Description & Specifications

Specifications

Chemical Name or Material Polyethylene
TSCA TSCA
Density 0.95g/mL
Name Note High density
Physical Form Granules
MDL Number MFCD00084423
CAS 9002-88-4
Merck Index 14,7567
Odor Odorless
Quantity 250g