Please login to your online account to display your discounted pricing

Alfa Aesar™ Polyethylene glycol succinate for glc

$74.05 - $969.74

Chemical Identifiers

Chemical Name or Material Polyethylene glycol succinate for glc
CAS 37340-09-3
Molecular Formula (CH2)2[CO2(CH2CH2O)nH]2
MDL Number MFCD00084421
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1172614

 
alfa aesar™
A11726-14
25g Each for $74.05

AAA1172622

 
alfa aesar™
A11726-22
100g Each for $241.30

AAA1172636

 
alfa aesar™
A11726-36
500g Each for $969.74
Description & Specifications

Specifications

Quantity 25g
Quantity 25g
Chemical Name or Material Polyethylene glycol succinate for glc
CAS 37340-09-3
Molecular Formula (CH2)2[CO2(CH2CH2O)nH]2
MDL Number MFCD00084421
TSCA TSCA