Please login to your online account to display your discounted pricing

Polyethylene glycol, MP Biomedicals™


Quantity 1kg

Manufacturer: mp biomedicals  0215191601

 View more versions of this product

Catalog No. ICN15191601

  • / Each
 

Description & Specifications

Chemical Identifiers

Density 1.1 to 1.2g/mL
Quantity 1kg
Chemical Name or Material Polyethylene glycol
CAS 25322-68-3
Physical Form White powder
Molecular Formula (C2H4O)x.H2O
Formula Weight 8000g/mol
Synonym Alkapol PEG-200

Specifications

Chemical Name or Material Polyethylene glycol
CAS 25322-68-3
Physical Form White powder
Molecular Formula (C2H4O)x.H2O
Formula Weight 8000g/mol
Synonym Alkapol PEG-200