Please login to your online account to display your discounted pricing

Polyethylene glycol, MP Biomedicals™

$118.29 - $118.29

Chemical Identifiers

Chemical Name or Material Polyethylene glycol
CAS 25322-68-3
Physical Form Powder
Molecular Formula (C2H4O)x.H2O
Formula Weight 8000
Synonym Alkapol PEG-200
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN19483901

 
mp biomedicals
19483901
1kg Each for $118.29
Description & Specifications

Specifications

Formula Weight 8000
Physical Form Powder
Color White
Quantity 1kg
Density 1.1 to 1.2g/mL
Chemical Name or Material Polyethylene glycol
Molecular Formula (C2H4O)x.H2O
CAS 25322-68-3
Synonym Alkapol PEG-200