missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Polyethylene glycol monomethylether, 550, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04156122
Description
Polyethylene glycol monomethylether, 550 is used for formulating screens and for optimization.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Colorless to Yellow | |
Viscous Liquid | |
18°C to 20°C | |
MFCD00084416 | |
methyl cellosolve, ethanol, 2-methoxy, ethylene glycol monomethyl ether, methyl oxitol, 2-methoxy-1-ethanol, methoxyethanol, 3-oxa-1-butanol, egme, monomethyl glycol, dowanol em | |
100 g | |
COCCO | |
76.10 | |
CHEBI:46790 |
Polyethylene glycol monomethylether, 550 | |
9004-74-4 | |
(C2H4O)nCH4O | |
1.10 g/mL | |
Slightly miscible with water. | |
XNWFRZJHXBZDAG-UHFFFAOYSA-N | |
2-methoxyethan-1-ol | |
8019 | |
>110°C (230°F) |
Chemical Identifiers
9004-74-4 | |
76.10 | |
XNWFRZJHXBZDAG-UHFFFAOYSA-N | |
8019 | |
COCCO |
(C2H4O)nCH4O | |
MFCD00084416 | |
methyl cellosolve, ethanol, 2-methoxy, ethylene glycol monomethyl ether, methyl oxitol, 2-methoxy-1-ethanol, methoxyethanol, 3-oxa-1-butanol, egme, monomethyl glycol, dowanol em | |
CHEBI:46790 |
Safety and Handling
RTECSNumber : PE8268000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only