missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Polyethylene glycol monomethylether, 1,900, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 04156336

 View more versions of this product

Catalog No. AA4156336


Description

Description

Polyethylene glycol monomethylether is used in the introduction of hydrophilic sites into polymers. It is also used to stabilize polymer emulsions. Further, it is employed in the synthesis of comb polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

White to Yellow
Solid Mass
52°C
MFCD00084416
Slightly miscible with water.
XNWFRZJHXBZDAG-UHFFFAOYSA-N
2-methoxyethanol
8019
>110°C (230°F)
Polyethylene glycol monomethylether 1,900
9004-74-4
(C2H4O)nCH4O
methyl cellosolve, ethanol, 2-methoxy, ethylene glycol monomethyl ether, methyl oxitol, 2-methoxy-1-ethanol, methoxyethanol, 3-oxa-1-butanol, egme, monomethyl glycol, dowanol em
500 g
COCCO
76.10
CHEBI:46790

Chemical Identifiers

9004-74-4
76.10
XNWFRZJHXBZDAG-UHFFFAOYSA-N
8019
2-methoxyethanol
(C2H4O)nCH4O
MFCD00084416
methyl cellosolve, ethanol, 2-methoxy, ethylene glycol monomethyl ether, methyl oxitol, 2-methoxy-1-ethanol, methoxyethanol, 3-oxa-1-butanol, egme, monomethyl glycol, dowanol em
CHEBI:46790
COCCO
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only