missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Polyethylene glycol monomethyl ether p-toluenesulfonate, M.W. 5,000

Manufacturer:  Alfa Aesar™ 4682403

 View more versions of this product

Catalog No. AA4682403



Specifications

Specifications

Polyethylene glycol monomethyl ether p-toluenesulfonate
58320-73-3
2-methoxyethyl p-toluenesulfonate, p-toluenesulfonic acid 2-methoxyethyl ester, 2-methoxyethyl tosylate, ethanol, 2-methoxy-, 1-4-methylbenzenesulfonate, 2-methoxyethyltosylate, acmc-209e3c, aronis23919, 2-methoxyethyl-p-toluenesulfonate, 2-methoxyethyl p-toluenesulphonate, ethylene glycol methyl ether tosylate
CC1=CC=C(C=C1)S(=O)(=O)OCCOC
230.278
Powder
1g
M.W. 5,000
C10H14O4S
TZXJJSAQSRHKCZ-UHFFFAOYSA-N
2-methoxyethyl 4-methylbenzenesulfonate
86977
White

Chemical Identifiers

58320-73-3
230.278
2-methoxyethyl p-toluenesulfonate, p-toluenesulfonic acid 2-methoxyethyl ester, 2-methoxyethyl tosylate, ethanol, 2-methoxy-, 1-4-methylbenzenesulfonate, 2-methoxyethyltosylate, acmc-209e3c, aronis23919, 2-methoxyethyl-p-toluenesulfonate, 2-methoxyethyl p-toluenesulphonate, ethylene glycol methyl ether tosylate
2-methoxyethyl 4-methylbenzenesulfonate
C10H14O4S
TZXJJSAQSRHKCZ-UHFFFAOYSA-N
86977
CC1=CC=C(C=C1)S(=O)(=O)OCCOC
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

SDS Documents