missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Polyethylene glycol monomethyl ether p-toluenesulfonate, M.W. 5,000, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04682406
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
White | |
M.W. 5,000 | |
58320-73-3 | |
2-methoxyethyl p-toluenesulfonate, p-toluenesulfonic acid 2-methoxyethyl ester, 2-methoxyethyl tosylate, ethanol, 2-methoxy-, 1-4-methylbenzenesulfonate, 2-methoxyethyltosylate, acmc-209e3c, aronis23919, 2-methoxyethyl-p-toluenesulfonate, 2-methoxyethyl p-toluenesulphonate, ethylene glycol methyl ether tosylate | |
TZXJJSAQSRHKCZ-UHFFFAOYSA-N | |
2-methoxyethyl 4-methylbenzenesulfonate | |
86977 |
Polyethylene glycol monomethyl ether p-toluenesulfonate | |
Powder | |
C10H14O4S | |
5 g | |
CC1=CC=C(C=C1)S(=O)(=O)OCCOC | |
230.278 |
Chemical Identifiers
58320-73-3 | |
230.278 | |
2-methoxyethyl p-toluenesulfonate, p-toluenesulfonic acid 2-methoxyethyl ester, 2-methoxyethyl tosylate, ethanol, 2-methoxy-, 1-4-methylbenzenesulfonate, 2-methoxyethyltosylate, acmc-209e3c, aronis23919, 2-methoxyethyl-p-toluenesulfonate, 2-methoxyethyl p-toluenesulphonate, ethylene glycol methyl ether tosylate | |
2-methoxyethyl 4-methylbenzenesulfonate |
C10H14O4S | |
TZXJJSAQSRHKCZ-UHFFFAOYSA-N | |
86977 | |
CC1=CC=C(C=C1)S(=O)(=O)OCCOC |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only