missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Polyethylene glycol monomethyl ether methanesulfonate, M.W. 5,000

Manufacturer:  Alfa Aesar™ 4645403

 View more versions of this product

Catalog No. AA4645403



Specifications

Specifications

Polyethylene glycol monomethyl ether methanesulfonate
175172-61-9
MFCD03100948
BCKAHDGFNHDQST-UHFFFAOYSA-N
2-methoxyethyl methanesulfonate
146293
White
1g
M.W. 5,000
C4H10O4S
methanesulfonic acid 2-methoxyethyl ester, ethanol, 2-methoxy-, methanesulfonate, 2-methoxyethanol methanesulfonate, 2-methoxyethyl mesylate, 2-methoxyethylmethanesulfonate, acmc-1c990, 2-methoxyethyl methane sulfonate, methanesulfonic acid2-methoxyethyl ester, ethanol, 2-methoxy-, 1-methanesulfonate, methanesulfonic acid 2-methoxy-ethyl ester
COCCOS(=O)(=O)C
154.18
Granular Solid
58°C to 61°C

Chemical Identifiers

175172-61-9
154.18
BCKAHDGFNHDQST-UHFFFAOYSA-N
146293
COCCOS(=O)(=O)C
C4H10O4S
MFCD03100948
methanesulfonic acid 2-methoxyethyl ester, ethanol, 2-methoxy-, methanesulfonate, 2-methoxyethanol methanesulfonate, 2-methoxyethyl mesylate, 2-methoxyethylmethanesulfonate, acmc-1c990, 2-methoxyethyl methane sulfonate, methanesulfonic acid2-methoxyethyl ester, ethanol, 2-methoxy-, 1-methanesulfonate, methanesulfonic acid 2-methoxy-ethyl ester
2-methoxyethyl methanesulfonate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents