missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Polyethylene glycol monomethyl ether methanesulfonate, M.W. 1,000, Thermo Scientific Chemicals
$156.63 - $485.77
Chemical Identifiers
CAS | 175172-61-9 |
---|---|
Molecular Formula | C4H10O4S |
Molecular Weight (g/mol) | 154.18 |
MDL Number | MFCD03100948 |
InChI Key | BCKAHDGFNHDQST-UHFFFAOYSA-N |
PubChem CID | 146293 |
IUPAC Name | 2-methoxyethyl methanesulfonate |
SMILES | COCCOS(=O)(=O)C |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
175172-61-9 | |
154.18 | |
BCKAHDGFNHDQST-UHFFFAOYSA-N | |
2-methoxyethyl methanesulfonate |
C4H10O4S | |
MFCD03100948 | |
146293 | |
COCCOS(=O)(=O)C |
Specifications
White | |
175172-61-9 | |
MFCD03100948 | |
1 g | |
COCCOS(=O)(=O)C | |
154.18 | |
Polyethylene glycol monomethyl ether methanesulfonate, M.W. 1,000 |
Granular Solid | |
C4H10O4S | |
methanesulfonic acid 2-methoxyethyl ester, ethanol, 2-methoxy-, methanesulfonate, 2-methoxyethanol methanesulfonate, 2-methoxyethyl mesylate, 2-methoxyethylmethanesulfonate, acmc-1c990, 2-methoxyethyl methane sulfonate, methanesulfonic acid2-methoxyethyl ester, ethanol, 2-methoxy-, 1-methanesulfonate, methanesulfonic acid 2-methoxy-ethyl ester | |
BCKAHDGFNHDQST-UHFFFAOYSA-N | |
2-methoxyethyl methanesulfonate | |
146293 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only