missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Polyethylene glycol methyl ether methacrylate, M.W. 1,000

Manufacturer:  Alfa Aesar™ 46991MD

 View more versions of this product

Catalog No. AA46991MD



Specifications

Specifications

Polyethylene glycol methyl ether methacrylate
26915-72-0
MFCD00241432
KRCGBOKYIUDIFY-UHFFFAOYSA-N
2-[2-[2-(2-methoxyethoxy)ethoxy]ethoxy]ethyl 2-methylprop-2-enoate
93675
White
250mg
M.W. 1,000
C13H24O6
mPEG-methacrylate
CC(=C)C(=O)OCCOCCOCCOCCOC
276.329
Granular Solid
33°C to 38°C

Chemical Identifiers

26915-72-0
276.329
KRCGBOKYIUDIFY-UHFFFAOYSA-N
93675
CC(=C)C(=O)OCCOCCOCCOCCOC
C13H24O6
MFCD00241432
mPEG-methacrylate
2-[2-[2-(2-methoxyethoxy)ethoxy]ethoxy]ethyl 2-methylprop-2-enoate
Safety and Handling

Safety and Handling

GHS H Statement
H318-H317-H335-H315
Causes serious eye damage.
May cause an allergic skin reaction.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

TSCA : TSCA

Recommended Storage : 2°C to 8°C

SDS Documents