missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Polyethylene glycol methyl ether acrylate, M.W. 1,000

Manufacturer:  Alfa Aesar™ 4653703

 View more versions of this product

Catalog No. AA4653703



Specifications

Specifications

Polyethylene glycol methyl ether acrylate
32171-39-4
MFCD00803685
HFCUBKYHMMPGBY-UHFFFAOYSA-N
2-methoxyethyl prop-2-enoate
18392
White
M.W. 1,000
C6H10O3
2-methoxyethyl acrylate, methoxyethyl acrylate, 2-methoxyethoxy acrylate, 2-propenoic acid, 2-methoxyethyl ester, methyl cellosolve acrylate, 2-methoxyethanol, acrylate, ethylene glycol monomethyl ether acrylate, acrylic acid, 2-methoxyethyl ester, ethylene glycol methyl ether acrylate, sipomer mca
COCCOC(=O)C=C
130.143
Granular Solid
1g

Chemical Identifiers

32171-39-4
130.143
HFCUBKYHMMPGBY-UHFFFAOYSA-N
18392
COCCOC(=O)C=C
C6H10O3
MFCD00803685
2-methoxyethyl acrylate, methoxyethyl acrylate, 2-methoxyethoxy acrylate, 2-propenoic acid, 2-methoxyethyl ester, methyl cellosolve acrylate, 2-methoxyethanol, acrylate, ethylene glycol monomethyl ether acrylate, acrylic acid, 2-methoxyethyl ester, ethylene glycol methyl ether acrylate, sipomer mca
2-methoxyethyl prop-2-enoate
Safety and Handling

Safety and Handling

TSCA : TSCA

Recommended Storage : 2°C to 8°C

Documents