missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Polyethylene glycol methyl ether acrylate, M.W. 1,000, Thermo Scientific Chemicals
$163.30 - $640.78
Chemical Identifiers
CAS | 32171-39-4 |
---|---|
Molecular Formula | (C2H4O)nC4H6O2 |
Molecular Weight (g/mol) | NaN |
MDL Number | MFCD00803685 |
InChI Key | HFCUBKYHMMPGBY-UHFFFAOYSA-N |
Synonym | 2-methoxyethyl acrylate, methoxyethyl acrylate, 2-methoxyethoxy acrylate, 2-propenoic acid, 2-methoxyethyl ester, methyl cellosolve acrylate, 2-methoxyethanol, acrylate, ethylene glycol monomethyl ether acrylate, acrylic acid, 2-methoxyethyl ester, ethylene glycol methyl ether acrylate, sipomer mca |
PubChem CID | 18392 |
IUPAC Name | 2-methoxyethyl prop-2-enoate |
SMILES | COCCOC(=O)C=C |
Description
Polyethylene glycol methyl ether acrylate is used as a raw material in chemical synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
32171-39-4 | |
NaN | |
HFCUBKYHMMPGBY-UHFFFAOYSA-N | |
18392 | |
COCCOC(=O)C=C |
(C2H4O)nC4H6O2 | |
MFCD00803685 | |
2-methoxyethyl acrylate, methoxyethyl acrylate, 2-methoxyethoxy acrylate, 2-propenoic acid, 2-methoxyethyl ester, methyl cellosolve acrylate, 2-methoxyethanol, acrylate, ethylene glycol monomethyl ether acrylate, acrylic acid, 2-methoxyethyl ester, ethylene glycol methyl ether acrylate, sipomer mca | |
2-methoxyethyl prop-2-enoate |
Specifications
White | |
32171-39-4 | |
MFCD00803685 | |
Soluble in water | |
HFCUBKYHMMPGBY-UHFFFAOYSA-N | |
2-methoxyethyl prop-2-enoate | |
18392 |
Granular Solid | |
(C2H4O)nC4H6O2 | |
2-methoxyethyl acrylate, methoxyethyl acrylate, 2-methoxyethoxy acrylate, 2-propenoic acid, 2-methoxyethyl ester, methyl cellosolve acrylate, 2-methoxyethanol, acrylate, ethylene glycol monomethyl ether acrylate, acrylic acid, 2-methoxyethyl ester, ethylene glycol methyl ether acrylate, sipomer mca | |
1 g | |
COCCOC(=O)C=C | |
NaN | |
Polyethylene glycol methyl ether acrylate, M.W. 1,000 |
Safety and Handling
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only