missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Polyethylene glycol methyl ether acrylate, M.W. 1,000

Manufacturer:  Alfa Aesar™ 4653706

 View more versions of this product

Catalog No. AA4653706



Specifications

Specifications

Polyethylene glycol methyl ether acrylate
32171-39-4
MFCD00803685
HFCUBKYHMMPGBY-UHFFFAOYSA-N
2-methoxyethyl prop-2-enoate
18392
White
M.W. 1,000
C6H10O3
mPEG-acrylate
COCCOC(=O)C=C
130.143
Granular Solid
5g

Chemical Identifiers

32171-39-4
130.143
HFCUBKYHMMPGBY-UHFFFAOYSA-N
18392
COCCOC(=O)C=C
C6H10O3
MFCD00803685
mPEG-acrylate
2-methoxyethyl prop-2-enoate
Safety and Handling

Safety and Handling

TSCA : TSCA

Recommended Storage : 2°C to 8°C

Documents