missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Polyethylene glycol-lithium acetate soln.

Common solution for protein and DNA isolation from yeast

Chemical Identifiers

Synonym PEG-LiOAc soltuion; PEG-lithium acetate solution
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ62213AK View Documents Alfa Aesar™
J62213AK
250mL N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

PEG-LiOAc soltuion; PEG-lithium acetate solution

Specifications

PEG-LiOAc soltuion; PEG-lithium acetate solution
Colorless
Polyethylene glycol-lithium acetate soln.
Liquid
250mL
Safety and Handling

Safety and Handling

TSCA : TSCA

Recommended Storage : 2° to 8°C