missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Polyethylene glycol dimethacrylate, M.W. 1,000

Manufacturer:  Alfa Aesar™ 4653403

 View more versions of this product

Catalog No. AA4653403



Specifications

Specifications

Polyethylene glycol dimethacrylate
25852-47-5
MFCD00081877
Granular Solid
White
M.W. 1,000
CH3(CH2=)COC(OCH2CH2)nOCOC(=CH2)CH3
PEG-dimethacrylate
Moisture & Light Sensitive
1g

Chemical Identifiers

25852-47-5
CH3(CH2=)COC(OCH2CH2)nOCOC(=CH2)CH3
MFCD00081877
PEG-dimethacrylate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Recommended Storage : −30°C to −10°C

SDS Documents