missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Polyethylene glycol diacrylate, M.W. 8,000

Manufacturer:  Alfa Aesar™ 46801MD

 View more versions of this product

Catalog No. AA46801MD



Specifications

Specifications

Polyethylene glycol diacrylate
26570-48-9
MFCD00081876
FCDZZFLRLVQGEH-UHFFFAOYSA-N
ethane-1,2-diol;prop-2-enoic acid
16212859
White
M.W. 8,000
C5H10O4
PEG-diacrylate
C=CC(=O)O.C(CO)O
134.131
Granular Solid
250mg

Chemical Identifiers

26570-48-9
134.131
FCDZZFLRLVQGEH-UHFFFAOYSA-N
16212859
C=CC(=O)O.C(CO)O
C5H10O4
MFCD00081876
PEG-diacrylate
ethane-1,2-diol;prop-2-enoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H318-H317-H335-H315
Causes serious eye damage.
May cause an allergic skin reaction.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

TSCA : TSCA

Recommended Storage : 2°C to 8°C

SDS Documents