Alfa Aesar™ Polyethylene glycol di-p-toluenesulfonate, M.W. 8,000
Manufacturer:  Alfa Aesar™ 4678706
 View more versions of this product
Catalog No. AA4678706
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications
Specifications
Polyethylene glycol di-p-toluenesulfonate
CH3C6H4SO2(OCH2CH2)nOSO2C6H4CH3
Granular Solid
5g
M.W. 8,000
PEG-di-p-tosylate
White
Chemical Identifiers
CH3C6H4SO2(OCH2CH2)nOSO2C6H4CH3
PEG-di-p-tosylate
Documents