missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Polyethylene glycol adipate for glc

$59.41 - $183.18

Chemical Identifiers

CAS 24938-37-2
Molecular Formula (CH2)4[CO2(CH2CH2O)nH]2
MDL Number MFCD00147998
Chemical Name or Material Polyethylene glycol adipate for glc
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1423814 Alfa Aesar™
A1423814
25g Each for $59.41
AAA1423822 Alfa Aesar™
A1423822
100g Each for $183.18
Specifications Description & Specifications

Chemical Identifiers

24938-37-2
(CH2)4[CO2(CH2CH2O)nH]2
MFCD00147998
Polyethylene glycol adipate for glc

Specifications

(CH2)4[CO2(CH2CH2O)nH]2
25g
Polyethylene glycol adipate for glc
24938-37-2
MFCD00147998

Safety and Handling

TSCA : TSCA