Please login to your online account to display your discounted pricing

Polyethylene Glycol 8000, Crystalline Powder, BAKER™, J.T.Baker™


A grade of chemicals for laboratory use

Quantity 2kg
Packaging Poly Bottle

Manufacturer: avantor performance  U22209

 View more versions of this product

Catalog No. 02003648

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 7000 to 9000 (Average)
Chemical Name or Material Polyethylene Glycol 8000
CAS 25322-68-3
Physical Form Crystalline Powder
Packaging Poly Bottle
Quantity 2kg

Specifications

Chemical Name or Material Polyethylene Glycol 8000
CAS 25322-68-3
Formula Weight 7000 to 9000 (Average)