Please login to your online account to display your discounted pricing

Polyethylene Glycol 400, Liquid, BAKER™, J.T.Baker™


A grade of chemicals for laboratory use

Quantity 19L
Packaging Poly Pail

Manufacturer: avantor performance  U21601

 View more versions of this product

Catalog No. 02003644

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 380 to 420 (Average)
Physical Form Liquid
Chemical Name or Material Polyethylene Glycol 400
CAS 25322-68-3
Specific Gravity 1L = 1.15kg
Packaging Poly Pail
Quantity 19L

Specifications

Chemical Name or Material Polyethylene Glycol 400
CAS 25322-68-3
Formula Weight 380 to 420 (Average)