missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Polyethylene Glycol 400, Liquid, BAKER™, J.T. Baker™
A grade of chemicals for laboratory use
Supplier: Avantor U21609
Specifications
Polyethylene Glycol 400 | |
25322-68-3 | |
MFCD01779601 | |
LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
ethane-1,2-diol | |
380 to 420 (Average) | |
1L = 1.15kg |
Liquid | |
(C2H4O)n | |
4 L | |
[H]OCCO | |
62.07 | |
Narrow Mouth Amber Glass Bottle |
Chemical Identifiers
25322-68-3 | |
62.07 | |
LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
[H]OCCO |
(C2H4O)n | |
MFCD01779601 | |
ethane-1,2-diol |