Please login to your online account to display your discounted pricing

Polyethylene Glycol 400, Liquid, BAKER™, J.T.Baker™


A grade of chemicals for laboratory use

Quantity 500mL
Packaging Narrow Mouth Amber Glass Bottle

Manufacturer: avantor performance  U21607

 View more versions of this product

Catalog No. 02003646

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 380 to 420 (Average)
Chemical Name or Material Polyethylene Glycol 400
CAS 25322-68-3
Physical Form Liquid
Quantity 500mL
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 1.15kg

Specifications

Chemical Name or Material Polyethylene Glycol 400
CAS 25322-68-3
Formula Weight 380 to 420 (Average)