Please login to your online account to display your discounted pricing

Polyethylene Glycol 3350, Powder, BAKER™, J.T.Baker™

A grade of chemicals for laboratory use

Quantity 1kg
Packaging Wide Mouth Amber Glass Bottle

Manufacturer: avantor performance  U221-08

 View all options for this product

Catalog No. 02003642

 

Description & Specifications

Chemical Identifiers

Packaging Wide Mouth Amber Glass Bottle
Quantity 1kg
Quantity 1kg
Chemical Name or Material Polyethylene Glycol 3350
CAS 25322-68-3
Purity Grade BAKER™
Physical Form Powder
Formula Weight Ave. Mol. Wt. 3000 to 3700

Specifications

Chemical Name or Material Polyethylene Glycol 3350
CAS 25322-68-3
Purity Grade BAKER™
Formula Weight Ave. Mol. Wt. 3000 to 3700