Please login to your online account to display your discounted pricing

Polyethylene glycol 200 Monooctylether, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  124750050

 View more versions of this product

Catalog No. AC124750050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Quantity 5g
Color Clear
TSCA TSCA
Density 0.9640g/mL
Refractive Index 1.4475 to 1.4495 (20°C, 589nm)
Health Hazard 3 GHS P Statement:
Keep container tightly closed.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Infrared Spectrum Authentic
CAS 27252-75-1
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May form explosive peroxides.
Chemical Name or Material Polyethyleneglycol 200 monooctylether
MDL Number MFCD00147977
Linear Formula CH3(CH2)7O(CH2CH2O)nHn=4-5

Specifications

Chemical Name or Material Polyethyleneglycol 200 monooctylether
CAS 27252-75-1
Linear Formula CH3(CH2)7O(CH2CH2O)nHn=4-5
MDL Number MFCD00147977
Accessories

GHS P Statement:
Keep container tightly closed.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May form explosive peroxides.

SDS & Safety Documents