Please login to your online account to display your discounted pricing

Polyethylene glycol 200 Monooctylether, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Polyethyleneglycol 200 monooctylether
CAS 27252-75-1
Linear Formula CH3(CH2)7O(CH2CH2O)nHn=4-5
MDL Number MFCD00147977
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC124750050

  SDS
thermo fisher scientific
124750050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Quantity 5g
Color Clear
TSCA TSCA
Density 0.9640g/mL
Refractive Index 1.4475 to 1.4495 (20°C, 589nm)
Health Hazard 3 GHS P Statement:
Keep container tightly closed.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Infrared Spectrum Authentic
CAS 27252-75-1
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May form explosive peroxides.
Chemical Name or Material Polyethyleneglycol 200 monooctylether
MDL Number MFCD00147977
Linear Formula CH3(CH2)7O(CH2CH2O)nHn=4-5
Safety and Handling

GHS P Statement:
Keep container tightly closed.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May form explosive peroxides.