Please login to your online account to display your discounted pricing

Polyethylene Glycol 1000, Waxy Soft Solid, BAKER™, J.T.Baker™


A grade of chemicals for laboratory use

Quantity 4L
Packaging Poly Bottle

Manufacturer: avantor performance  U21809

 View more versions of this product

Catalog No. 02003635

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 950 to 1050 (Average)
Chemical Name or Material Polyethylene Glycol 1000
CAS 25322-68-3
Physical Form Waxy Soft Solid
Packaging Poly Bottle
Specific Gravity 1L = 1.15kg
Quantity 4L

Specifications

Chemical Name or Material Polyethylene Glycol 1000
CAS 25322-68-3
Formula Weight 950 to 1050 (Average)