Please login to your online account to display your discounted pricing

Polyethylene Glycol 1000, Waxy Soft Solid, BAKER™, J.T.Baker™

A grade of chemicals for laboratory use

$179.41 - $1,639.57

Chemical Identifiers

Chemical Name or Material Polyethylene Glycol 1000
CAS 25322-68-3
Formula Weight 950 to 1050 (Average)
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003635

 
avantor performance
U21809
Poly Bottle 4L Each for $1,639.57

02003636

 
avantor performance
U21807
Poly Bottle 500mL Each for $179.41
Description & Specifications

Specifications

Formula Weight 950 to 1050 (Average)
Chemical Name or Material Polyethylene Glycol 1000
CAS 25322-68-3
Physical Form Waxy Soft Solid
Packaging Poly Bottle
Specific Gravity 1L = 1.15kg
Quantity 4L