Learn More
Polyethylene glycol 10,000, Thermo Scientific Chemicals
$73.93 - $154.32
Chemical Identifiers
CAS | 25322-68-3 |
---|---|
Molecular Formula | (C2H4O)n |
Molecular Weight (g/mol) | 62.07 |
MDL Number | MFCD01779601 |
InChI Key | LYCAIKOWRPUZTN-UHFFFAOYSA-N |
IUPAC Name | ethane-1,2-diol |
SMILES | [H]OCCO |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2195530
|
Thermo Scientific Chemicals
B2195530 |
250 g |
Each for $73.93
|
|
|||||
AAB219550B
|
Thermo Scientific Chemicals
B219550B |
1000 g |
Each for $154.32
|
|
|||||
Description
Polyethylene glycol is used to modify therapeutic proteins and peptides to increase their solubility. In gas chromatography, it is used as a polar stationary phase. It is also useful in preserving objects obtained from the ocean. As a dispersant, it is used in toothpastes. It finds application in blood banking as a potentiator, which is used to detect antigens and antibodies.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
25322-68-3 | |
62.07 | |
LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
[H]OCCO |
(C2H4O)n | |
MFCD01779601 | |
ethane-1,2-diol |
Specifications
Powder | |
58°C to 63°C | |
MFCD01779601 | |
Soluble in organic solvents,water and aromatic hydrocarbons. Slightly soluble in aliphatic hydrocarbons. | |
LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
ethane-1,2-diol | |
≈10,000 | |
Polyethylene glycol 10, 000 |
25322-68-3 | |
(C2H4O)n | |
1.2 | |
250 g | |
[H]OCCO | |
62.07 | |
265°C (509°F) |
Safety and Handling
EINECSNumber : 203-473-3
RTECSNumber : TQ4110000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only