missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Polyethylene glycol 1,500, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A162410B
Description
Cell fusion induced by polyethylene glycol(PEG) has become a standard method in somatic cell genetics. PEG promoted cell fusion is involved in the production of hybridoma cells.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Polyethylene glycol 1,500 | |
25322-68-3 | |
(C2H4O)n | |
14,7568 | |
Soluble in water and organic solvents. Insoluble in aliphatic hydrocarbons. | |
LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
ethane-1,2-diol | |
Odorless |
5 to 7 | |
44°C to 48°C | |
MFCD01779601 | |
PEG 1500 | |
1000 g | |
[H]OCCO | |
62.07 |
Chemical Identifiers
25322-68-3 | |
62.07 | |
LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
ethane-1,2-diol |
(C2H4O)n | |
MFCD01779601 | |
PEG 1500 | |
[H]OCCO |
Safety and Handling
EINECSNumber : 500-038-2
RTECSNumber : TQ4030000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only