Alfa Aesar™ Polyethylene Glove Bags
For handling air and moisture sensitive, high purity chemicals and materials
Manufacturer:  Alfa Aesar™ 92717LL
 View more versions of this product
Catalog No. AA92717LL
Description
Description
  • With inflatable chamber
Specifications
Specifications
68 x 43 x 30cm; 56cm entrance
4/bag
For handling air, moisture sensitive, high purity chemicals and materials
Documents