missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dynalon™ Polyethylene Dropping Pipets

Disposable polyethylene droppers with tapered tips and bellows-type bulbs.

Manufacturer:  Dynalon™ DAL262175

 View more versions of this product

Catalog No. S18751



Description

Description

  • Made of non-wetting material
  • Give excellent drop formation
  • Deliver 30 to 35 drops/mL
  • Easy to clean and extra safe

Specifications

Specifications

0.05 oz.
0.835 in.
5.38 x 0.835 in. (13.66 x 2.12cm)
Yes
13.66cm
Dropping Pipet
LDPE (Low-density Polyethylene)
0.05 oz., 1.5mL
2.12cm
Yes
5.38 in.
Tapered
1.5mL
Documents