Please login to your online account to display your discounted pricing

Dynalon™ Polyethylene Dropping Bottles


Easy to fill bottle — remove closure.

Manufacturer: dynalon™  87000103

 View more versions of this product

Catalog No. S31712

  • / Pack of 10 Each
  • / Case of 6 Pack
 

Description & Specifications

Specifications

Capacity 2.02 oz., 60mL
Capacity (Metric) 60mL
Capacity (English) 2.02 oz.
Includes LPDE dropping pipet
Material LDPE
Mouth Narrow

  • Narrow-mouth, low-density polyethylene bottle with LDPE dropping pipet
  • Bellows-type dropping pipet is fixed to closure