missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Polyethylene, chlorinated

Manufacturer:  Alfa Aesar™ 4414630

 View more versions of this product

Catalog No. AA4414630



Specifications

Specifications

Polyethylene
64754-90-1
Soluble in cyclohexanone and toluene
1.22
250g
Chlorinated
MFCD00084424
Powder
White

Chemical Identifiers

64754-90-1
MFCD00084424
Safety and Handling

Safety and Handling

RTECSNumber : KM6893750

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents