Please login to your online account to display your discounted pricing

Alfa Aesar™ Polyethylene, chlorinated

$72.32 - $204.92

Chemical Identifiers

Chemical Name or Material Polyethylene
CAS 64754-90-1
Name Note Chlorinated
MDL Number MFCD00084424
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4414630

  SDS
alfa aesar™
4414630
250g Each for $72.32

AA44146A1

  SDS
alfa aesar™
44146A1
1kg Each for $204.92
Description & Specifications

Specifications

Chemical Name or Material Polyethylene
TSCA TSCA
Name Note Chlorinated
Physical Form Granular
MDL Number MFCD00084424
CAS 64754-90-1
Color Off-white
Quantity 250g
RTECS Number KM6893750