Please login to your online account to display your discounted pricing

Polyethylene Acids and Corrosives Cabinets

Superior resistance to harmful vapors and spills from aggressive acids such as sulfuric, hydrochloric and nitric.

$2,411.10 - $2,411.10

Specifications

Type Storage Cabinet
Door Style Manual Close
For Use With (Application) Flammables
Includes 2 in. Dual Poly vents with caps, 4-poly leveling legs and specific warning labels
Material Polyethylene
View More Specs
Products
Catalog Number Mfr. No. Capacity (English) No. of Shelves Color Price Quantity    

S98125

 
fisher science education
CRAP22W_EMD
22 gal. 2 White Each for $2,411.10
Description & Specifications

Specifications

Type Storage Cabinet
Door Style Manual Close
For Use With (Application) Flammables
Includes 2 in. Dual Poly vents with caps, 4-poly leveling legs and specific warning labels
Material Polyethylene
Surface Coating Smooth

  • 100% Polyethylene construction with smooth, easy-to-clean finish
  • Outfitted with dual 2 in. (5.1cm) capped vents for controlling vapor buildup and migration
  • Internal and top sump capacities of at least 1 gal. (3.8L)
  • Polyethylene shelves adjust to maximize storage space
  • Storage for smaller containers inside doors expands usable space
  • Lockable manual doors bear “CAUTION CORROSIVES” label; have raised drip edge to keep spills in cabinet
  • 4 and 22 gal. (15.1 and 83.3L) models have recessed front base and center partition that divides interior into separate compartments
  • 44 gal. (166.5L) models have an up/down configuration for storage of noncompatible chemicals