missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Polyester acrylate/methacrylate, PEAM-1769

Recommended for use as a base resin or an additive to reduce stress

Manufacturer:  Alfa Aesar™ 4635114

 View more versions of this product

Catalog No. AA4635114



Specifications

Specifications

Polyester acrylate/methacrylate
921214-61-1
Amber
PEAM-1769
Liquid
25g

Chemical Identifiers

921214-61-1
Safety and Handling

Safety and Handling

GHS H Statement
H361-H315-H319-H335
Suspected of damaging fertility or the unborn child.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P281-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Recommended Storage : 2°C to 8°C

SDS Documents