missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 770, Thermo Scientific Chemicals
$50.21 - $364.84
Chemical Identifiers
CAS | 9016-00-6 |
---|---|
Molecular Formula | (C2H6OSi)n |
Molecular Weight (g/mol) | 74.15 |
MDL Number | MFCD00084411 |
InChI Key | SEUDSDUUJXTXSV-UHFFFAOYSA-N |
IUPAC Name | dimethylsilanone |
SMILES | C[Si](C)(-*)O-* |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA42497AD
|
Thermo Scientific Chemicals
042497AD |
50 mL |
Each for $50.21
|
|
|||||
AA42497AK
|
Thermo Scientific Chemicals
042497AK |
250 mL |
Each for $138.89
|
|
|||||
AA42497K2
|
Thermo Scientific Chemicals
042497K2 |
1 L |
Each for $364.84
|
|
|||||
Description
Polydimethylsiloxane, trimethylsiloxy is used in coating Auxiliary Agents. It is also used as Pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
9016-00-6 | |
74.15 | |
SEUDSDUUJXTXSV-UHFFFAOYSA-N | |
C[Si](C)(-*)O-* |
(C2H6OSi)n | |
MFCD00084411 | |
dimethylsilanone |
Specifications
Colorless | |
9016-00-6 | |
-40°C | |
MFCD00084411 | |
Soluble in aromatic hydrocarbon,partially soluble in aliphatic and insoluble in water. | |
SEUDSDUUJXTXSV-UHFFFAOYSA-N | |
dimethylsilanone | |
135°C (275°F) | |
1.397 |
Liquid | |
>205°C | |
(C2H6OSi)n | |
0.918 g/mL | |
50 mL | |
C[Si](C)(-*)O-* | |
74.15 | |
Odorless | |
Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 770 |
Safety and Handling
RTECSNumber : TQ2690000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only